Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbamazepine ≥98%

Supplier: Enzo Life Sciences
Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

550-084-G005
89152-134EA 67.6 USD InStock
89152-134
Carbamazepine ≥98%
Carbamazepine
Anticonvulsant and analgesic

  • Formulation: White to off-white crystalline solid.

Anticonvulsant and analgesic.


Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
MDL Number: MFCD00005073
CAS Number: 298-46-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/03/45390803.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty