Would you like to log in again?
BIO-PROBE® Herpes Simplex Virus Probe, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-40838
89165-726EA 369.08 USD InStock
89165-726
BIO-PROBE® Herpes Simplex Virus Probe, Enzo Life Sciences
The Herpes Simplex Virus (HSV) BioProbe® labeled probe is prepared by nick translation of two clones of HSV DNA, one 8 kb fragment from HSV 1 DNA and one 16 kb fragment from HSV 2 DNA.
The probe is specific for herpes simplex viruses. It does not cross hybridize to cytomegalovirus nor Epstein-Barr virus, but is capable of detecting clinical isolates of HSV. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty