Would you like to log in again?
BIOSCORE® Screening And Amplification Kit (20 Reactions), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42440
89165-748EA 815.23 USD InStock
89165-748
BIOSCORE® Screening And Amplification Kit (20 Reactions), Enzo Life Sciences
Nucleic Acid Reagents Isothermal Amplification Reagents
The BIOSCORE® Screening and amplification kit is a rapid and reliable method to determine the quality of genomic DNA isolated from formalin-fixed paraffin-embedded (FFPE) tissue samples before use in array-based genome-wide copy number analysis and other genomic profiling applications.
This dual purpose kit utilizes a whole genome amplification (WGA) method to identify genomic DNA samples that are suitable for microarray analysis prior to labeling and to predict sample performance with virtually 100% concordance. After prescreening, the amplified genomic DNA can then be used in genomic profiling assays such as array analysis.
Delivery information: The Kit includes Primers (400 µL), Enzyme (20 µL), Stop Buffer (110 µL), Nucleotide Mix (200 µL) and Nuclease-free Water (1 mL).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty