Would you like to log in again?
BIGGY Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C5160 C5161 C5163 C5162 C5161
89405-158EA 5023.03 USD InStock
CA89405-154 89405-158 89405-156 89405-154 89405-152
BIGGY Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ BiGGY Agar (Bismuth Sulfite Glucose Glycine Yeast Agar) is a selective and differential medium used in the isolation and presumptive identification of Candida spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty