Would you like to log in again?
Blot Module, Cytiva
Supplier: Cytiva
80-6418-96 80-6418-96
95044-746EA 667.35 USD InStock
95044-746 CA95044-746L
Blot Module, Cytiva
Blotting Apparatus
Perform 1-D and 2-D electrophoresis and electrotransfer in a single compact unit.
  • Complete semi-wet blotting in 45 minutes with only 300 mL buffer
  • Choose precast gels from a variety of manufacturers
  • Perform two second-dimension separations using 7 cm IPG strips in 15 hours
Cast, run, and blot mini-gels in four streamlined components: Gel module, blot module, lower buffer chamber, safety lid.
Two mini-gels or four mini-blots at once, using 10 × 10.5 cm gel cassettes for increased separation distances.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty