Would you like to log in again?
MMP-1 Human, Cytiva
Supplier: Cytiva
RPN2610 RPN2610
95019-108EA 1274.92 USD InStock
CA95019-108L 95019-108
MMP-1 Human, Cytiva
The MMP-1 Human Biotrak ELISA system has been specifically designed for research purposes.
  • Suitable for use with serum, plasma (EDTA, heparin, citrate) and cell culture supernatant samples
  • Biotrak ELISAs measure MMP protein levels in human serum, plasma, and cell culture samples
MMP-1 may be measured in the range 6.25 to 100 ng/ml. The assay is based on a two site ELISA ‘sandwich’ format. Standards and samples are incubated in microplate wells pre-coated with anti-MMP-1 antibody. Any MMP-1 present will be bound to the wells, other components of the sample being removed by washing and aspiration.
A second polyclonal antibody to MMP-1 is then bound to the MMP-1. Any excess is removed by washing and aspiration. The second antibody bound to the wells is detected using donkey anti-rabbit horseradish peroxidase. The amount of peroxidase bound to each well is determined by the addition of TMB ‘ready-to-use’ substrate.
The reaction is stopped by addition of an acid solution, and the resultant color read at 450 nm in a microplate spectrophotometer. The concentration of MMP-1 in a sample is determined by interpolation from a standard curve. Each pack contains reagent for 96 determinations. This allows the construction of a standard curve plus the measurement of 42 samples in duplicate.
Ordering information: Kit contents include Microplate: 12×8 well strips coated with mouse anti-MMP-1; Assay buffer 1: 10 ml of phosphate buffer concentrate; Assay buffer 2: 10 ml of phosphate buffer concentrate; Standard: 1 ml human MMP-1 (recombinant); Antibody: lyophilised rabbit anti-MMP-1; Peroxidase conjugate: lyophilised donkey anti-rabbit horseradish peroxidase; Wash buffer: 12.5 ml phosphate buffer concentrate; TMB substrate: 3,3’,5,5’-Tetramethylbenzidine (TMB)/hydrogen peroxide.
Delivery information: The assay recognises total human MMP-1, ie. free MMP-1 and that complexed with inhibitors such as TIMP-1. It does not recognise MMP-1 bound by the non-specific protease inhibitor α2-macroglobulin. There is no cross-reactivity with α2-macroglobulin alone. It does not recognise MMP-1 from other species as far as we are aware. There is no significant cross-reactivity or interference in the assay with MMP-3, 2 and 9. Pro MMP-1, and MMP-1 complexed with TIMP-1 cross-react in the assay. The cross-reactivity with TIMP-1 and its lack of interference in the presence of MMP-1 standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty