Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Silver Enhancing Kit, MP Biomedicals
Supplier: MP Biomedicals
08678211
IC678211PK 579.13 USD InStock
IC678211
Silver Enhancing Kit, MP Biomedicals
Blotting Kits
For Immunoblotting This ImmuMark™ silver enhancing kit is used for amplifying the signal from gold particles attached to proteins or antigens immobilized on membranes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty