Would you like to log in again?
ProteoChrom® HPTLC Plates for Peptid Analysis, MilliporeSigma
Supplier: MilliporeSigma
1.05651.0001 1.05651.0001 1.05650.0001 1.05650.0001
EM1.05650.0001EA 760.59 USD InStock
CA1.05651.0001 EM1.05650.0001 CA1.05650.0001 EM1.05651.0001
ProteoChrom® HPTLC Plates for Peptid Analysis, MilliporeSigma
TLC Plates
We've enhanced our ProteoChrom® plates to make separation highly efficient - especially when peptides and protein digests need to be analyzed
ProteoChrom® plates take performance to new levels. Applications are highly reproducible thanks to optimized separation and staining procedures. With extra-thin 100 µm layers, the plates are incredibly sensitive. They're also perfect for use with aqueous solvent systems since they prove highly stable in water. For added convenience, our ProteoChrom® plates include easy-to-read and detailed protocols.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty