Would you like to log in again?
CC70-2 Field Carrying Case and Standards Kit for the IQ340/350, Hach
Supplier: Hach
CC70-2
82026-938EA 137.6 USD InStock
82026-938
CC70-2 Field Carrying Case and Standards Kit for the IQ340/350, Hach
Carriers Bottle Carriers/Containers
CC70-2 Field Carrying Case and Standards Kit for the IQ340/350
Contains Conductivity Standards Bottles 100 mL each of 100 uS, 1000 uS, 10,000 uS, 53 mS and Conductivity Test sample bottle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty