Would you like to log in again?
HQ411d Benchtop Meter Package with PHC201 Gel pH Probe, Hach
Supplier: Hach
8507560
89500-878EA 3576.44 USD InStock
89500-878
HQ411d Benchtop Meter Package with PHC201 Gel pH Probe, Hach
Electrochemical Meters pH and Multi-Parameter Meters
The Hach HQ411d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements - IntelliCAL™ smart probes store all calibrations in the probe
  • Ultra-bright screen and large font size
  • Internal USB ports
  • Convenient kit includes everything you need to start testing
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate. The sealed PHC201 pH electrode is an ideal general-purpose probe for use in multiple measurement applications.
Ordering information: Includes HQ411D laboratory pH meter, universal electrode stand with electrode holder, PHC201 standard gel filled pH electrode with 1 m cable, pH buffer kit 4,01/7,00/10,01 (500 mL), pH electrode storage solution (500 mL), 4 alkaline 1,5V AA batteries, power supply adapter, power cord, and operating documentation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty