Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Standard Line Sterile Centrifuge Tubes with Flat Caps, Conical-Bottom, PP

Supplier: VWR
VWR®
CTNA305 CTNA304
10026-078CS 224.37 USD InStock
10026-078 10026-076
VWR® Standard Line Sterile Centrifuge Tubes with Flat Caps, Conical-Bottom, PP
Tubes Centrifuge Tubes
Disposable conical-bottom centrifuge tubes are made of high-quality, USP Class 6 gamma-resistent polypropylene.

  • Available in two volumes: 15 and 50 ml
  • Longer length screw caps with sealing ring prevent leakage
  • Large, unerasable frosted white writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube
  • Max. rotation speed up to 7500×g, autoclavable at 121 °C and freezable to –80 °C
  • Leak-proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters

Tubes feature black printed graduations and smooth inner walls for easy filling and sample preparation. A white printed writing area on the side allows for permanent coding. Tubes are sterilized by irradiation. Tubes and caps are autoclavable and freezable. RNase-free, DNase-free and non-pyrogenic.

Easy-to-read black graduations are accurate to ±2%, with 1 ml increments (15 ml) or 2.5 ml increments (50 ml).

Certifications: ISO 13485:2003 and ISO 9001:2008.

Ordering information: Bagged tubes are packaged in durable and resealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability. All tubes are packaged with caps attached and sterilized with irradiation. Plastic racks are available as optional accessaries in seven colors. An assorted color pack offers one of each color: blue, green, orange, red, and yellow. Racks accommodate plug or flat bottom tubes.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty