Would you like to log in again?
SP Bel-Art Smooth, Chisel-Type Rigid Scraper, Bel-Art Products, a part of SP
Supplier: Bel-Art
H36819-0000
13197-402EA 58.06 USD InStock
13197-402
SP Bel-Art Smooth, Chisel-Type Rigid Scraper, Bel-Art Products, a part of SP
Scrapers
This scraper has a flat polystyrene blade, 5.9 cm (2⅓”) wide, with a tapered, sharp edge that is slightly curved.
  • Polystyrene construction
  • Tapered, sharp end edge
  • Rounded side edges
  • Suited for contact with foods, drugs, and cosmetics
The side edges are rounded and the blade is 10.8 cm (4¼”) long. Made of FDA-grade materials for direct contact with foods, drugs, and cosmetics.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty