Would you like to log in again?
Film Cleaner, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74317
100492-138EA 19.75 USD InStock
100492-138
Film Cleaner, Electron Microscopy Sciences
Film Processing Supplies
It cleans positives and negatives
It removes tape-marks, grease, pencil, ink, dirt, smudges, and fingerprints. Leaves no residue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty