Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Kodak Electron Image Film SO-163, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
KODAK®
74144 74145 74142
102091-270CS 5638.13 USD InStock
102091-270 102091-268 102091-264
Kodak Electron Image Film SO-163, Electron Microscopy Sciences
Photographic Film
  • Fine grain, blue sensitive emulsion.
  • 7mil ESTAR base.
  • Slightly faster than Electron Image Plates.
  • Twice as fast as EM film 4489.
  • May use anti-fog.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty