Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
LABCOM SW sensION+ GLP Software, Hach
Supplier: Hach
LZW8999.99
89233-224EA 1370 USD InStock
89233-224
LABCOM SW sensION+ GLP Software, Hach
Software Laboratory Software
LabCom PC software, for bi-directional communication of sensION+ GLP instruments with a PC.
  • Convenient PC tool for data management and instrument control
Complete management of the instrument from the PC. Programming and configuration of calibrations and measurements from the PC. Start calibrations and measurements from the PC. Automate the measurements of up to 4 instruments with sampler. On line graph view of measured value. Results table personalization, the user selects the displayed information. Storage of calibration and measuring results in a data base where the data can be viewed or printed at any moment. Print report of individual sample or results summary for several samples. Data export to Excel.
Delivery information: Kit including CD, cable with 9-contact connector and USB adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty