Would you like to log in again?
GeneMate Neon Microcentrifuge Tubes
Supplier: Scientific Specialties
1210-29-210IS
490003-246BX 135.79 USD InStock
490003-246
GeneMate Neon Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
BioExpress offers brightly colored microtubes for those researchers with a need for easy identification and color coding of samples and experiments
Flat cap tubes are graduated and fully autoclavable. These tubes are not recommended for boiling applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty