Would you like to log in again?
GeneMate Clearview Co-Polymer 1.7 ml Graduated Microcentrifuge Tubes with High Chemical Resistance, Scientific Specialties
Supplier: Scientific Specialties
1220-05 1220-00-210IS
490003-261BX 104.68 USD InStock
490003-261 490003-250
GeneMate Clearview Co-Polymer 1.7 ml Graduated Microcentrifuge Tubes with High Chemical Resistance, Scientific Specialties
Tubes Microcentrifuge Tubes
  • Easy to open
  • Ultimate in clarity, glass-like
  • Positive seal design (-90˚C to 120˚C)
  • Chamfered tube opening for easy & rapid cap closure
  • Rated to 20,000 x g
  • Limited boiling applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty