Would you like to log in again?
Freezer Box and Cell Dividers, 15 and 50 ml, Custom Biogenic Systems
Supplier: Custom Biogenic Systems
B3CL D16L D36L
490006-586EA 7.8 USD InStock
490006-586 490006-590 490006-572
Freezer Box and Cell Dividers, 15 and 50 ml, Custom Biogenic Systems
Boxes Cryoboxes
For upright freezer and chest freezer racks
Box Dimensions: 143 x 143 x 73 mm (5.63 x 5.63 x 2.88") Divider's Dimensions: 138 x 138 x 70 mm (5.44 x 5.44 x 2.75") Cell Opening for Cell Divider for 50 mL Tubes: 35 x 35 mm (1.38 x 1.38") Cell Opening for the Cell Divider for 15 mL Tubes: 22 x 22 mm (0.88 x 0.88")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty