Would you like to log in again?
HyperSep 96-Well Plate Manifold, Thermo Scientific
Supplier: Thermo Scientific
60103-351
10047-010EA 3802.75 USD InStock
10047-010
HyperSep 96-Well Plate Manifold, Thermo Scientific
Manifolds
Provides processing of 96-well SPE, SLE, Protein precipitation and Lab/Filter plates
This format provides higher throughput processing of samples allowing for higher efficency workflows and reproducible results.
Delivery information: 96-Well Plate Vacuum Manifold includes base, lid, waist collection tray and vacuum gauge.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty