Would you like to log in again?
HyperSep Universal Vacuum Manifold, Thermo Scientific
Supplier: Thermo Scientific
60104-230
10047-012EA 5926.7 USD InStock
10047-012
HyperSep Universal Vacuum Manifold, Thermo Scientific
Manifolds
Allows processing of SPE and SLE cartridges (3,4 flangeless and 6 mL) or 96-well plates
Allows flexibility in sample processing.
Delivery information: Includes manifold, base/gauge, flask and stopper, tubing and spigots.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty