Would you like to log in again?
Paddle Scrapers, 52" without Holes, Metal Detectable, Remco Products
Supplier: Remco
6777MD2 6777MD3 6777MD4 6777MD5
76008-334EA 232.35 USD InStock
76008-334 76008-328 76008-330 76008-332
Paddle Scrapers, 52" without Holes, Metal Detectable, Remco Products
Scrapers
This two-in-one tool is handy for both mixing and scraping, and is great for a variety of applications such as soup, confectionery, dough, and sauce manufacturing as well as many other food processing or other mixing tasks
One-piece construction and the unique long-handle design make it easy to use for many jobs. The semi-ferrous additive in this paddle allows for use in conjunction with a metal detectable system. Metal detectable plastic is a lightweight and less costly alternative to stainless steel mixing paddle scrapers, and will reduce fatigue for the user. Polypropylene and semi-ferrous additive materials.
Certifications: FDA compliant raw material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty