Would you like to log in again?
SP Bel-Art Dry-Keeper™ Plus Auto-Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art
H42061-0000
24982-200EA 3297.01 USD InStock
24982-200
SP Bel-Art Dry-Keeper™ Plus Auto-Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
White ABS desiccator cabinet has vertical profile and automatic 120VAC (15 watt) electronic dehumidification system that maintains relative humidity levels at 12 to 20% (cabinet interior rH% levels can differ depending upon room temperature, room rH% and usage conditions) and keeps dry air circulating throughout the cabinet.
  • White ABS construction
  • Vertical profile
  • 2-hour absorption period
  • One-handed opening with simple latch design
It features a 2-hour absorption period with a ½ hour desiccant regeneration cycle and the permanent desiccant is contained in the control unit.
Two perforated shelves rest on preset rail locations with a maximum capacity three shelves (additional shelf sold separately). A large, in-door dial hygrometer provides easy visualization of rH%. The clear door opens to the right exposing an interior opening measuring 25 x 43 cm (10 x 17"). A one-touch latch permits opening and closing of door using one hand for easier access. The 1.2 cu. ft. interior volume cabinet is stackable to conserve space.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty