Would you like to log in again?
What kind of bacteria is it?
Supplier: Avantor
WARD470045-032
470140-300KT 266.29 USD InStock
470140-300 470045-032
What kind of bacteria is it?
Educational Classroom Kits and Activities
In this lab activity, students identify an unknown bacterial sample using gross examination, cell morphology, and biochemical tests with Becton-Dickenson EnteroPluri Tubes. 
  • Identification Tests Used in Real-World Laboratories
  • Kit Employs NGSS Life Science DCI LS1
  • Pre and Post-Laboratory Assessments Included
  • Estimated Lab Time: Two Lab Periods, 45 Minutes Each
  • Assessments and Vocabulary Words Target Common Core Standards
This kit is aligned with all published National Standards. Pre and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core standards.
The teacher guide contains specific “Did You Know” concepts that link the real world with the scientific standards explored.
Ordering information: This kit includes 5 slides of Gram-stained mixed bacteria, 5 color-reaction key cards/EnteroPluri score sheets, 1 CCIS (EnteroPluri interpretation guide), 5 EnteroPluri tubes, and 1 coupon redeemable for 5 plates of bacteria to be used as "unknown" samples. A compound microscope, a 100x oil-immersion lens, and an incubator are needed but not included. These products contain living or perishable materials. Please specify a delivery date. Bacterial and fungal cultures are restricted and can be ordered only by educational and research professionals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty