Would you like to log in again?
Vikan® Aluminimun Light-Duty Handle, 51", Remco Products
Supplier: Vikan
2945
76006-954EA 12.93 USD InStock
76006-954
Vikan® Aluminimun Light-Duty Handle, 51", Remco Products
Brooms
This standard broom handle can be used with all brooms, squeegees and scrapers.
  • Aluminum handle with polypropylene grip
  • Euro (hygienic) thread type
The handle with the attached item should reach to the chin of the user to prevent occupational back injuries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty