Would you like to log in again?
Rotary Motion Motor Kit
Supplier: VERNIER SOFTWARE & TECH SE
MK-RMV
470113-966EA 11.99 USD InStock
470113-966
Rotary Motion Motor Kit
Tachometers
This kit includes a small electric motor with pulley, rubber band belt, motor clip, and mounting screw. The motor can be attached to the Rotary Motion Sensor, and be used as an analog tachometer, or generator. You can also perform experiments that investigate motor efficiency in different conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty