Would you like to log in again?
Large Vane Thermo-Anemometer Series, Extech
Supplier: Extech
AN320-NISTL AN320 AN310 AN300-NIST AN310-NIST AN300
10111-872EA 1108.37 USD InStock
10111-872 10171-820 10171-822 10111-868 10171-824 10111-870
Large Vane Thermo-Anemometer Series, Extech
Anemometers
Extech’s 4" (10 cm) Large Vane Thermo-Anemometer series allows for more precise readings on larger size ducts
All units feature a built-in thermistor for air temperature measurements. Psychrometer models also feature wet bulb (-4 to 140°F or -20 to 60°C), relative humidity measurement (0.1 to 99.9%RH) and dew point measurement (23 to 140°F or -5 to 60°C).
Each meter simultaneously displays Airflow or Air Velocity and Temperature
Certifications: Psychrometer plus CO2 models feature wet bulb (-4 to 140°F or -20 to 60°C), relative humidity measurement (0.1 to 99.9%RH), dew point measurement (23 to 140°F or -5 to 60°C) and CO2 measurement (23 to 140°F or -5 to 60°C).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty