Would you like to log in again?
Refrigerant Leakage Detector, Extech
Supplier: Extech
RD300
10068-320EA 429.19 USD InStock
10068-320
Refrigerant Leakage Detector, Extech
Leak Detection Systems Gas Leak Detection Systems
Extech’s Refrigerant Leakage Detector uses a heated diode sensor
It is easy to use and is ideal for detecting leakages from air-conditioning units and cooling systems that use all standard refrigerants. The multi-colored LED light bar indicates the level of leaking refrigerant. The built-in, bright LED convenience light located at the sensor tip illuminates dimly lit inspection areas.
Certifications: CE
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty