Would you like to log in again?
Large Capacity Condensers
Supplier: DWK Life Sciences
458000-2950
KT458000-2950EA 1209.92 USD InStock
KT458000-2950
Large Capacity Condensers
Condensers
This large capacity, highly efficient condenser is ideal for pre-pilot and pilot plant operations
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Full length Standard Taper 29/42 joints; standard taper outer joint at the top, standard taper inner drip joint at the bottom.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty