Would you like to log in again?
Aerator for BOD Bottles, WHEATON, DWK Life Sciences
Supplier: DWK Life Sciences
227700
16171-450EA 219.74 USD InStock
16171-450
Aerator for BOD Bottles, WHEATON, DWK Life Sciences
Tubes Gas Inlet Tubes
Manufactured from WHEATON 200 low extractable borosilicate glass.
  • 40 to 60 µm porosity glass frit
  • EPA Method 245.1 Manual Cold Vapor Technique
Used in determining inorganic forms of mercury and organic mercurials in drinking, surface and saline waters, as well as domestic and industrial waste.
Certifications: Conforms to USP Type I and also ASTM E 438 Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty