Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pimamicin, milky suspension

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Natamycin

0215625420
IC15625420EA 384.27 USD InStock
IC15625420
Pimamicin, milky suspension
Pimaricin
Pimaricin is a saline suspension with approximately 2.5% pimaricin.
Pimaricin is an amphoteric macrolide antifungal antibiotic from Streptomyces natalensis or S. chattanoogensis.

  • Concentration: 2.5% aqueous suspension
  • Presentation: Milky Suspension
  • Storage temperature: Store at + 4 °C. Protect from light.

It is used for a variety of fungal infections, mainly topically. It is used as an ergosterol and cholesterol binding agent to study lipid bilayer dynamics, especially in fungal cells. It is used to study pimaricin biosynthesis1 and as a fungicide in agar media.

An antifungal polyene macrolide that binds specifically to ergosterol and blocks fungal growth. However, unlike nysatin and filipin, pimaricin does not change the permeability of the plasma membrane.


Formula: C₃₃H₄₇NO₁₃
MW: 665.733 g/mol
Density: 1,020 g/mL (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00135085
CAS Number: 7681-93-8
Order Now
Specification Test Results
Identity Test Passes

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty