Would you like to log in again?
True RMS Power Clamp Meter, Extech
Supplier: Extech
CM83-NIST CM82 CM82-NIST CM85-NIST CM85 CM83
10748-340EA 1143.54 USD InStock
10748-340 10748-338 10748-336 10748-314 10748-312 10748-310
True RMS Power Clamp Meter, Extech
Electrical Meters Voltmeters
FLIR offers an excellent choice of industrial-grade Power Clamp Meters engineered with advanced power analysis and variable frequency drive filtering functions required by electrical troubleshooters
These meters are perfect for the electrician who needs a safe, capable combination tool.
Harmonics: 1st-25th Order
CE, UL.
Ordering information: Meters come with CAT IV Silicone Test Leads, 6 AAA Batteries, User Manual/CD and Limited Lifetime Warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty