Would you like to log in again?
VWR® Mini Logger-Trac™ Temperature Datalogger
Supplier: VWR
6548
10510-926EA 75.16 USD InStock
10510-926
VWR® Mini Logger-Trac™ Temperature Datalogger
Data Loggers
Perfect solution for monitoring material in transit as well as reagents, clinical samples, vaccines, pharmaceuticals and food items in any location.
  • Reusable datalogger with great storage capacity
  • Waterproof design
  • Cumulative alarm duration timer for extra safety with exportable data
  • Integrates with MaxiThermal software for complete data analysis
Waterproof design (IP66) eliminates concerns when located in damp environments or areas that are required to be washed down.
Know the Temperature Excursion Duration immediately - Active LCD display technology allows the duration (cumulative time) for the temperature excursion to be viewed on the display immediately without the need to download data. The user may select the acceptable temperature parameters (high and low temperature values) in 1° increments. The cumulative alarm duration timer is started each time the reading is outside of the acceptable temperature parameters to provide a total excursion time since the start of recording.
Reusable/reprogrammable datalogger with a storage capacity of 7680 readings. Active LCD Display shows the following important data current temperature, minimum/maximum temperatures, recording status, run time, alarm (excursion) cumulative time, alarm tally.
Ordering information: Accessories not included. For complete data analysis and reprogramming of unit, order the MaxiThermal Software and cable/cradle (for additional MaxiThermal Software details, please see Accessories for VWR® Traceable® Logger-Trac™ Temperature Datalogger). Contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty