Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Antibody (Cy3®)

Supplier: Boster
BA1031
10208-962EA 556.27 USD InStock
10208-962
Anti-IgG Antibody (Cy3®)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3®
Epitope:
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty