Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
S150 High Power Ultrasonic Cleaner, Elma
Supplier: Elma
1071279
10724-496EA 4696.47 USD InStock
10724-496
S150 High Power Ultrasonic Cleaner, Elma
Ultrasonic Cleaners
The powerful Elmasonic S150 ultrasonic bath is designed to process large numbers of small or large laboratory samples, with an internal tank depth of less than 4 inches. The tank configuration is also optimum for cleaning surgical instruments.
  • Specially designed for sample preparation and medical instrument cleaning
  • Large capacity shallow tank
  • High intensity ultrasonic power
  • 3 modes optimized for mixing, cleaning, and degassing
The S150 operates at 37 kHz in three ultrasonic modes: Normal mode for mixing, Sweep mode for cleaning, or Degas mode. The timer can be set from 1-30 minutes or continuous operation. A series of LEDs continuously and simultaneously show set time and remaining time.
The microprocessor-controlled generator in this sonic cleaner maintains constant ultrasonic power, sensing and adjusting for light or heavy loads in the tank. Digital operation with power tracking ensures reproducible performance from run to run. The rear drain is controlled by a valve on the side of the tank for easy access. For safety purposes the unit turns off automatically after 12 hours of continuous operation. The tank cover can be flipped over and used as a drip tray.
Dependable, state-of-the-art Elma ultrasonic cleaners are made in Germany.
Certifications: CE, ISO9001, ISO13485.
Ordering information: The cover is included. The basket is available and is ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty