Would you like to log in again?
Avantor® Seradigm, Iron Supplemented Bovine Calf Serum (BCS)
Supplier: Avantor
Avantor® Seradigm
2100-050
10158-360EA 6.98 USD InStock
10158-360
Avantor® Seradigm, Iron Supplemented Bovine Calf Serum (BCS)
Sera (Serum)
Iron Supplemented Bovine Calf Serum is sourced from US origin animals, aged up to six months and raised on a diet consisting mainly of formula.
  • 100% US Origin
  • Cost effective alternative to FBS
  • Similar performance to FBS in many established cell lines
  • ISIA Certified Supply Chain for Traceability
These source animals have naturally high levels of transferrin
When the serum is supplemented with iron, the iron binds to the unbound transferrin, resulting in a product with three to four times the amount of available iron compared to FBS or equine serum. This product is an excellent, cost-effective alternative to FBS and often performs as well as FBS in many established cell lines. In some applications, it even out-performs FBS and equine serum.
Avantor Seradigm is a fully-integrated supplier of animal sera that provides the cell culture community with access to the most reliable supply of high performance, exceptional quality Fetal Bovine Serum (FBS) and cost-effective FBS alternatives. Our approach to sourcing and manufacturing is refreshingly unique and distinct from the competition, resulting in product performance and quality unlike any other.
Certifications: ISIA certified supply chain for traceability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty