Would you like to log in again?
Mighty Small Multiple Gel Caster, Cytiva
Supplier: Cytiva
80-6142-51
CA95044-774
Mighty Small Multiple Gel Caster, Cytiva
Blotting Kits
To cast 10 x 8 cm glass plates.
Ordering information: Includes 20 x Glass plates, rectangular (10 x 8 cm); 10 x Aluminium plates, notched (10 x 8 cm); 2 x Clamps; 100 x Wax paper sheets; Space saver plates; 5 x Filler sheets; Filler plug set; Space-mate assembly template. Combs and spacers need to be ordered separately. Recommended gradient maker: SG 100 (code no 80-6196-09).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty