Would you like to log in again?
Amersham™ Hybond™ SEQ Western Blotting Membrane, 0.2 PVDF, Cytiva
Supplier: Cytiva
Amersham™ Hybond™
10600030 10600030
10120-026PK 823.19 USD InStock
CA10061-210 10120-026
Amersham™ Hybond™ SEQ Western Blotting Membrane, 0.2 PVDF, Cytiva
Transfer Membranes
0.2 µm pore size for higher surface area, resulting in better binding of low molecular weight proteins. Amersham Hybond SEQ 0.2 PVDF is a 0.2 µm pore size hydrophobic membrane designed specifically for protein sequencing applications. The small pore size of this membrane minimizes ‘blow-through’ and increases protein binding over a wide range of molecular weights.
  • For Western blotting
  • High protein binding capacity for easy signal detection
  • Chemical resistance needed for N-terminal sequencing
  • High protein retention even after harsh washing steps
  • Efficient capture of proteins during transfer minimizes sample loss
  • Available in popular precut sizes for your application
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty