Would you like to log in again?
DALTsix Gel Caster, Cytiva
Supplier: Cytiva
80648546 80-6485-46
10147-856EA 2307.46 USD InStock
CA11028-386 10147-856
DALTsix Gel Caster, Cytiva
Blotting Kits
DALTsix Gel Caster holds up to six 1.0 or 1.5-mm cassettes with separator sheets for casting homogeneous or gradient gels
DIGE approved
If fewer gels are to be cast, the unused volume is filled with blank cassette inserts and filler sheets.
Ordering information: Includes 7 x Separator Sheets 0.5 mm and 6 x Filler Sheets 1.0 mm. For gradient gel casting, a pump and gradient maker are needed and have to be ordered separately. Cassettes and glass plates have to be ordered separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty