Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Cored JRT Combo Unit Bathroom Tissue Dispenser, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
09551 09551
10571-338EA 165.32 USD InStock
10571-338 CA10672-500
Cored JRT Combo Unit Bathroom Tissue Dispenser, KIMBERLY-CLARK PROFESSIONAL®
Wipe Dispensers
The Cored JRT Combo Unit Bathroom Tissue Dispenser are made of durable grey plastic with smoked transparent cover for use with either two full 9.38" diameter rolls or one standard 13" diameter roll plus stub roll.
Rolls have a 3.8" width and a 3.25" diameter core. With two full 9.38" rolls, it holds more than the equivalent in length of ten standard rolls. Design minimizes run-out, waste and maintenance time.
Hinged front cover, push button for easy opening or common key lock to reduce pilferage and tear-off bars on sides and front of dispenser opening. Easy-to-use mechanism to meet the customer's needs. Used with or without a key.
Certifications: 1994 ADA Standards for Accessible Design, 28 CFR Part 36 - Local rules may vary.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty