Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acridine orange (chlorid salt), certified by the Biological Stain Commission nucleus stain (DNA/RNA) C.I. 46005

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Basic Orange 143,6-bis(Dimethylamino)acridine, HClAO

10050
100503-634EA 194.06 USD InStock
100503-634
Acridine orange (chlorid salt), certified by the Biological Stain Commission nucleus stain (DNA/RNA) C.I. 46005
Acridine orange (chlorid salt)
Used for Pickett's fluorescence method for fungi fluorescent against a dark background and for differentiation of nucleic acids and cytology.

Ultrastructural probes on DNA templates within human bone marrow and lymph node cells (Frenstar, J., Adv. Cell Mol. Biol., 3, 1(1974)).
Can be used to detect changes in DNA template activity within cells (Frenster, J. Cell Nucl., 1, 565 (1974)).
Can be used as a fluorescent dye for cytochemistry (J. Histochem. Cytochem., 31, 737 ((1983) J. Lab. Med., 15 (3),180, (1984)).

Certifications: Certified by the Biological Stain Commission.

Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.


Formula: C₁₇H₁₉N₃·HCl·xH₂O
MW: 301.81 g/mol (anhy)
MDL Number: MFCD00012660
CAS Number: 65-61-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty