Would you like to log in again?
Flame Resistant DC1 Contractor Coveralls with Max Comfort™ Material, Gray, MCR Safety
Supplier: MCR Safety
Max Comfort™
DC1G48T DC1G52 DC1G52T DC1G60T DC1G46T DC1G58T DC1G50 DC1G50T DC1G56T DC1G62T DC1G58 DC1G64T DC1G54T DC1G46 DC1G44 DC1G38T DC1G54 DC1G60 DC1G66T DC1G56 DC1G36 DC1G36T
10812-884EA 188.92 USD InStock
10812-884 10812-894 10812-886 10812-896 10812-874 10812-888 10812-910 10812-876 10812-898 10812-854 10812-900 10812-856 10812-902 10812-904 10812-906 10812-908 10812-880 10812-890 10812-882 10812-860 10812-892 10812-870
Flame Resistant DC1 Contractor Coveralls with Max Comfort™ Material, Gray, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
  • Comfortable 88% cotton 12% Nylon blend
  • Two needle reinforced stitching on major seams for durability
  • Heavy-duty two-way YKK® brass break-away zipper with Neomex® tape
  • Multiple pockets on chest, back, hips, arm and leg
  • NFPA 2112, NFPA 70E, UL Certified
  • Lifetime FR guarantee
The DC1 coveralls feature a comfortable 88% cotton/12% nylon twill material that meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty