Would you like to log in again?
Flame Resistant DC1 Contractor Coveralls with Max Comfort™ Material, Tan, MCR Safety
Supplier: MCR Safety
Max Comfort™
DC1T66T DC1T46 DC1T56 DC1T56T DC1T50T DC1T36T DC1T40T DC1T62T DC1T36 DC1T42T DC1T48T DC1T54 DC1T60 DC1T52T DC1T54T DC1T38 DC1T58T DC1T64T DC1T44T DC1T46T DC1T60T DC1T34 DC1T50 DC1T38T DC1T58
10813-116EA 188.92 USD InStock
10813-116 10813-112 10813-286 10813-264 10813-120 10813-284 10813-282 10813-280 10813-108 10813-106 10813-302 10813-104 10813-268 10813-300 10813-266 10813-288 10813-276 10813-298 10813-110 10813-296 10813-274 10813-272 10813-294 10813-292 10813-290
Flame Resistant DC1 Contractor Coveralls with Max Comfort™ Material, Tan, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
  • Three needle reinforced stitching on major seams for durability
  • Heavy duty two-way YKK® brass break-away zipper with Nomex® tape
  • Protective storm flap with snap closure to cover front zipper
  • Two chest pockets with mitered flaps, snap closure, plus pen pocket
  • Two front hip pockets on coverall pants
  • Side pass through with snap closure
  • Two back patch pockets – left pocket with flap and snap closure
  • Bi-swing action back gussets for ease of movement and comfort
  • Pen pocket on left arm
  • Tool pocket on right leg
  • Adjustable snap closure on cuffs of sleeves and legs
  • All snaps have reinforcement strips
The DC1 coveralls feature a comfortable 88% cotton/12% nylon twill material that meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty