The VWR® Standard Series Manual Defrost Laboratory Series Freezers are designed specifically with general-purpose applications in mind.
- Adjustable operating temperature range: –15 to –25 °C (5 to –13 °F)
- Direct-cool fixed evaporator shelves
These units are ideal when stable temperatures averaging -20°C are required but precise temperature control, maintenance, and alarms are not necessary. These freezers feature a manual defrost design to eliminate damaging temperature spikes from defrost cycles. This ensures valuable sample security. Defrosting of the unit must be performed periodically to prevent severe ice build-up.
Freezer 10819-410 comes with leveling legs (not casters). An optional roller base platform is available (97043-518).
Certifications: UL/C-UL listed.
Ordering information: Two-year parts and labor warranty, five years on compressor parts.