Would you like to log in again?
Medline Sharps Container, Essendant
Supplier: Essendant
MIIMDS705153H
102830-236EA 7.28 USD InStock
102830-236
Medline Sharps Container, Essendant
Bins Sharps Containers
Sharps medical waste disposal system.
  • Lid design limits access to container contents and facilitates one-handed operation
  • Translucent lid provides easy-to-see fill levels
  • Large openings accommodate larger syringes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty