Would you like to log in again?
Kuglen Iris Hook and Lens Manipulator, OR Grade, Sklar
Supplier: Sklar
66-7726 66-7725 66-7727
10198-100EA 125.95 USD InStock
10198-100 10198-102 10198-104
Kuglen Iris Hook and Lens Manipulator, OR Grade, Sklar
Retractors
The Sklar Kuglen iris hook and lens manipulator is used primarily in cataract surgeries.
  • Made of stainless steel
The specialized tip of this hook allows the surgeon to utilize a push-pull pattern where the iris tissue and lens material can be pulled back, or retracted, and also pushed back into proper alignment as necessary.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty