Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Septa Cap for Vials, C&G Containers
Supplier: C&G Containers
CAH242005I04
71000-006EA 0.88 USD InStock
71000-006
Septa Cap for Vials, C&G Containers
Closures Chromatography Vial Closures
Septa caps are not processsed, pre-cleaned, nor certified.
Pack or case of vial closures do not contain any documentation, such as Certificate of Analysis or Cleaning Protocol Form. Septa caps are designed for use with C&G glass vials, all of which use a 24-414 closure.
Septa caps are polyprolylene open-top caps with a PTFE septa ultrasonically bonded (not glued). These are one-piece caps. The PTFE septa is a 3.1mm fluoropolymer resin/silicone septa laminate (overall thickness 0.125", ±0.006", silicone thickness is 0.115", PTFE is 0.010").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty