Would you like to log in again?
Neptune 70-118 Kevlar® Sleeve, Ansell
Supplier: Ansell
Neptune
222146
80080-585CS 499.39 USD InStock
80080-585
Neptune 70-118 Kevlar® Sleeve, Ansell
Workwear
Kevlar® cut- and heat-resistant sleeve.
  • Double-layered construction for extra cut resistance, with good comfort and flexibility
  • Provides forearm protection from minor heat and burn injuries
  • Thumb slot version prevents the sleeve from twisting while in use
  • Ambidextrous
Use in the automotive, glass, metal fabrication, and durable goods industries. Uses include glass handling, metal handling, and assembly.
Certifications: ANSI Level 4 Cut Protection; ANSI Level 3 Abrasion Protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty