Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.

Silicone oil, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses

Synonyms: Polyphenyl-methylsiloxane

AC16385-0010 AC16385-0025 AC16385-5000
103516-528EA 230 USD InStock
103516-528 200056-346 200056-344
Silicone oil, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
Silicone oil
  • Refractive index: 1.403 - 1.406


Melting Pt: -50 °C
Density: 0.96
Flash Pt: 300 °C
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty