Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.

Oil, for heat transfer, Thermo Scientific

Supplier: Thermo Scientific
AC26877-0025 AC26877-0100
103509-948EA 285 USD InStock
103509-948 200056-078
Oil, for heat transfer, Thermo Scientific
Circulator Bath Fluids
MDL No.: MFCD00147787
Density: 0.86
Melting Point: -6°C
Flash Point: 230°C
Appearance: Clear light yellow Oily liquid
Storage Temperature: Ambient

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty