Would you like to log in again?
MATra-si Reagent, PromoKine
Supplier: PromoKine
PK-CT-2021-005 PK-CT-2021-100 PK-CT-2021-020
10181-202EA 107.37 USD InStock
10181-202 10181-206 10181-204
MATra-si Reagent, PromoKine
Transfection Reagents
MATra-siRNA contains specifically coated magnetic nanoparticles, which are optimized to effectively bind and stabilize siRNA.
  • Provides optimal delivery of siRNA into many different cell types
  • Enables efficient and prolonged gene silencing
  • Uses magnet-assisted transfection (MATra) technology
MATra-siRNA contains specifically coated magnetic nanoparticles, which are optimized to effectively bind and stabilize siRNA. It provides optimal delivery of siRNA into many different cell types enabling efficient and prolonged gene silencing using the Magnet-assisted transfection (MATra) technology.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty